CAS RN 3158-56-3
CAS RN 3158-56-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Pentadecylresorcinol, TRC
CAS: 3158-56-3 Molecular Formula: C21H36O2 Molecular Weight (g/mol): 320.51 Synonym: 5-Pentadecyl-1,3-benzenediol,3-n-Pentadecyl-5-hydroxyphenol,5-(n-Pentadecyl)resorcinol,Adipostatin A,Hydrogenated Cardol,NSC 776 IUPAC Name: 5-pentadecylbenzene-1,3-diol SMILES: O=C(Nc1cc(c(Cl)cc1)Oc2c(Cl)cc(cn2)C(F)(F)F)NC(=O)c3c(F)cccc3F