missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furfuraldazine, TRC

Product Code. 30134014
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134014 1 g 1g
30004969 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134014 Supplier Toronto Research Chemicals Supplier No. TRCN4938301G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112537-96-9
Molecular Formula C10 H6 N4 O6
Molecular Weight (g/mol) 278.178
Synonym 5-Nitro-2-furancarboxaldehyde (E,E)-(5-Nitro-2-furanyl)methylene]hydrazone, USP Nitrofurazone Related Compound A, Nitrofurazone USP Related Compound A, (E,E)-N,N'-Bis[(5-nitrofuran-2-yl)methylidene]hydrazine, 2-Furancarboxaldehyde, 5-nitro-, [(5-nitro-2-furanyl)methylene]hydrazone, (E,E)- (9CI), (E,E)-N,N'-Bis[(5-nitrofuran-2-yl)methylidene]hydrazine (5-Nitrofurfural Azine)
IUPAC Name (E)-1-(5-nitro-2-furyl)-N-[(E)-(5-nitro-2-furyl)methyleneamino]methanimine
SMILES [O-][N+](=O)c1oc(\C=N\N=C\c2oc(cc2)[N+](=O)[O-])cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H6N4O6/c15-13(16)9-3-1-7(19-9)5-11-12-6-8-2-4-10(20-8)14(17)18/h1-6H/b11-5+,12-6+
Formula Weight 278.0287
Percent Purity >95%
Chemical Name or Material 5-Nitro-2-furfuraldazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.