CAS RN 112537-96-9
CAS RN 112537-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Nitro-2-furfuraldazine, TRC
CAS: 112537-96-9 Molecular Formula: C10 H6 N4 O6 Molecular Weight (g/mol): 278.178 Synonym: 5-Nitro-2-furancarboxaldehyde (E,E)-(5-Nitro-2-furanyl)methylene]hydrazone,USP Nitrofurazone Related Compound A,Nitrofurazone USP Related Compound A,(E,E)-N,N'-Bis[(5-nitrofuran-2-yl)methylidene]hydrazine,2-Furancarboxaldehyde, 5-nitro-, [(5-nitro-2-furanyl)methylene]hydrazone, (E,E)- (9CI),(E,E)-N,N'-Bis[(5-nitrofuran-2-yl)methylidene]hydrazine (5-Nitrofurfural Azine) IUPAC Name: (E)-1-(5-nitro-2-furyl)-N-[(E)-(5-nitro-2-furyl)methyleneamino]methanimine SMILES: [O-][N+](=O)c1oc(\C=N\N=C\c2oc(cc2)[N+](=O)[O-])cc1