missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhydantoin, 98%

Product Code. 11368545
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368545 1 g 1g
11378545 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368545 Supplier Thermo Scientific Alfa Aesar Supplier No. L11992.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 616-03-5 | C4H6N2O2 | 114.104 g/mol

5-Methylhydantoin is usede as a reactant for preparation of N-chlorohydantoin, Mitsunobu reaction, Pseudomonas putida strain allowing conversion into L-amino acids, D-hydantoinase from Agrobacteria tumefaceins and enzymatic racemization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylhydantoin is usede as a reactant for preparation of N-chlorohydantoin, Mitsunobu reaction, Pseudomonas putida strain allowing conversion into L-amino acids, D-hydantoinase from Agrobacteria tumefaceins and enzymatic racemization.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-03-5
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.104
MDL Number MFCD00022398
InChI Key VMAQYKGITHDWKL-UHFFFAOYSA-N
Synonym 5-methylhydantoin, hydantoin, 5-methyl, 2,4-imidazolidinedione, 5-methyl, 5-methyl-2,4-imidazolidinedione, d-5-methylhydantoin, 5-methyl-hydantoin, spectrum_001906, 2, 5-methyl, spectrum3_000916, spectrum4_001200
PubChem CID 69216
IUPAC Name 5-methylimidazolidine-2,4-dione
SMILES CC1C(=O)NC(=O)N1

Specifications

Melting Point 150°C to 152°C
Quantity 1 g
Beilstein 81081
Solubility Information Insoluble in water.
Formula Weight 114.1
Percent Purity 98%
Chemical Name or Material 5-Methylhydantoin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.