accessibility menu, dialog, popup

UserName

CH3NHOHNO

CAS RN 616-03-5

IUPAC Name: 5-methylimidazolidine-2,4-dione
Synonyms: methyl hydantoin
Molecular Weight (g/mol): 114.1
Molecular Formula: C4H6N2O2
InChi Key: VMAQYKGITHDWKL-UHFFFAOYNA-N
SMILES: CC1NC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results