Learn More
5-Formylpyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Product Code. 15467737
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467737 1 g 1g
15477737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15467737 Supplier Thermo Scientific Alfa Aesar Supplier No. H37151.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 848093-29-8
Molecular Formula C12H16BNO3
Molecular Weight (g/mol) 233.074
MDL Number MFCD04038759
InChI Key WVIMCYHOZKNOCS-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinaldehyde, 5-formylpyridine-3-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbaldehyde, 3-formyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carboxaldehyde, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinecarboxaldehyde, abbypharma ap-11-5936, 5-formylpyridin-3-boronic acid pinacol ester
PubChem CID 4992640
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbaldehyde
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)C=O
Specifications
Quantity 1 g
Formula Weight 233.08
Percent Purity 97%
Chemical Name or Material 5-Formylpyridine-3-boronic acid pinacol ester