accessibility menu, dialog, popup

UserName

CAS RN 848093-29-8

BOOH3CCH3H3CH3CNO

CAS RN 848093-29-8

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbaldehyde
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 233.07
Molecular Formula: C12H16BNO3
InChi Key: WVIMCYHOZKNOCS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C=O)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results