Learn More
5-Formyl-5,6,7,8-tetrahydropteroic Acid Hydrochloride Salt, TRC
Product Code. 30138287
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138287 100 mg 100mg
30010538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138287 Supplier Toronto Research Chemicals Supplier No. TRCF700980100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2469007-14-3
Molecular Formula C15H17ClN6O4
Molecular Weight (g/mol) 380.79
Synonym 4-[[(2-Amino-5-formyl-1,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoic Acid, 5-Formyltetrahydropteroic Acid Hydrochloride Salt, p-[[(2-Amino-5-formyl-5,6,7,8-tetrahydro-4-hydroxy-6-pteridinyl)methyl]amino]benzoic Acid Hydrochloride Salt
IUPAC Name 4-(((2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzoic acid
SMILES O=C1C2=C(NCC(CNC3=CC=C(C(O)=O)C=C3)N2C=O)NC(N)=N1.Cl
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H16N6O4.ClH/c16-15-19-12-11(13(23)20-15)21(7-22)10(6-18-12)5-17-9-3-1-8(2-4-9)14(24)25;/h1-4,7,10,17H,5-6H2,(H,24,25)(H4,16,18,19,20,23);1H
Formula Weight 380.1
Percent Purity >95%
Chemical Name or Material 5-Formyl-5,6,7,8-tetrahydropteroic Acid Hydrochloride Salt
Grade HPLC