CAS RN 2469007-14-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Formyl-5,6,7,8-tetrahydropteroic Acid Hydrochloride Salt, TRC
CAS: 2469007-14-3 Molecular Formula: C15H17ClN6O4 Molecular Weight (g/mol): 380.79 Synonym: 4-[[(2-Amino-5-formyl-1,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl)methyl]amino]benzoic Acid,5-Formyltetrahydropteroic Acid Hydrochloride Salt,p-[[(2-Amino-5-formyl-5,6,7,8-tetrahydro-4-hydroxy-6-pteridinyl)methyl]amino]benzoic Acid Hydrochloride Salt IUPAC Name: 4-(((2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzoic acid SMILES: O=C1C2=C(NCC(CNC3=CC=C(C(O)=O)C=C3)N2C=O)NC(N)=N1.Cl