Learn More
5-Ethoxy-2-mercaptobenzimidazole, 97%, Thermo Scientific™
Product Code. 11374755
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374755 5 g 5g
11384755 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11374755 Supplier Thermo Scientific Alfa Aesar Supplier No. L17781.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55489-15-1
Molecular Formula C9H10N2OS
Molecular Weight (g/mol) 194.252
MDL Number MFCD01326487
InChI Key WUSCBOFBIYZVCQ-UHFFFAOYSA-N
Synonym 5-ethoxy-2-mercaptobenzimidazole, 5-ethoxy-1h-benzo d imidazole-2-thiol, 5-ethoxy-1h-benzimidazole-2-thiol, 5-ethoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-ethoxybenzimidazole, 5-ethoxybenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-ethoxy-1,3-dihydro, acmc-1arr4, ksc603o0j, 5-ethoxy-2-benzimidazolethiol
PubChem CID 790793
IUPAC Name 5-ethoxy-1,3-dihydrobenzimidazole-2-thione
SMILES CCOC1=CC2=C(C=C1)NC(=S)N2
Specifications
Melting Point 246°C to 250°C
Quantity 5 g
Beilstein 4310013
Formula Weight 194.26
Percent Purity 97%
Chemical Name or Material 5-Ethoxy-2-mercaptobenzimidazole