accessibility menu, dialog, popup

UserName

CAS RN 55489-15-1

CH3OHNSHN

CAS RN 55489-15-1

IUPAC Name: 5-ethoxy-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5-ethoxy-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 194.25
Molecular Formula: C9H10N2OS
InChi Key: WUSCBOFBIYZVCQ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2NC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results