Learn More
5-Chloro Acesulfame, TRC
Product Code. 30135970
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135970 25 mg 25mg
30135347 250 mg 250mg
30007566 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135970 Supplier Toronto Research Chemicals Supplier No. TRCC36325025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 72827-08-8
Molecular Formula C4 H4 Cl N O4 S
Molecular Weight (g/mol) 197.6
Synonym 5-Chloro-6-methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide, Acesulfam Potassium Imp. B (EP)
IUPAC Name 5-chloro-6-methyl-2,2-dioxooxathiazin-4-one
SMILES CC1=C(Cl)C(=O)NS(=O)(=O)O1
Spezifikation
Quantity 25 mg
InChI Formula InChI=1S/C4H4ClNO4S/c1-2-3(5)4(7)6-11(8,9)10-2/h1H3,(H,6,7)
Formula Weight 196.955
Percent Purity >95%
Chemical Name or Material 5-chloro-6-methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide
Grade HPLC