CAS RN 72827-08-8
CAS RN 72827-08-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Chloro Acesulfame, TRC
CAS: 72827-08-8 Molecular Formula: C4 H4 Cl N O4 S Molecular Weight (g/mol): 197.6 Synonym: 5-Chloro-6-methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide,Acesulfam Potassium Imp. B (EP) IUPAC Name: 5-chloro-6-methyl-2,2-dioxooxathiazin-4-one SMILES: CC1=C(Cl)C(=O)NS(=O)(=O)O1