missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro Acesulfame, TRC

Product Code. 30135347
Change view
Click to view available options
Quantity:
10 mg
25 mg
250 mg
Unit Size:
10mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135347 250 mg 250mg
30135970 25 mg 25mg
30007566 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135347 Supplier Toronto Research Chemicals Supplier No. TRCC363250250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72827-08-8
Molecular Formula C4 H4 Cl N O4 S
Molecular Weight (g/mol) 197.6
Synonym 5-Chloro-6-methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide, Acesulfam Potassium Imp. B (EP)
IUPAC Name 5-chloro-6-methyl-2,2-dioxooxathiazin-4-one
SMILES CC1=C(Cl)C(=O)NS(=O)(=O)O1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H4ClNO4S/c1-2-3(5)4(7)6-11(8,9)10-2/h1H3,(H,6,7)
Formula Weight 196.955
Percent Purity >95%
Chemical Name or Material 5-chloro-6-methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.