missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-7047612
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378876 1 g 1g
11388876 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378876 Supplier Thermo Scientific Alfa Aesar Supplier No. L19514.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-49-1
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD03094221
InChI Key OZKAHSNNKADHTK-UHFFFAOYSA-N
Synonym 5-chloro-2-fluorobenzene-1-sulfonyl chloride, 5-chloro-2-fluorobenzenesulphonyl chloride, 5-chloro-2-fluoro-benzenesulfonyl chloride, 5-chloro-2-fluorobenzensulphonyl chloride, pubchem5094, 5-chloro-2-fluorobenzenesulfonylchloride, acmc-1ctc7, chloro 5-chloro-2-fluorophenyl sulfone, 5-chloro-2-fluorobenzenesulphonylchloride, 5-chloro-2-fluorophenylsulfonyl chloride
PubChem CID 2778273
IUPAC Name 5-chloro-2-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Cl)S(=O)(=O)Cl)F

Specifications

Melting Point 41°C to 44°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 229.06
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluorobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.