accessibility menu, dialog, popup

UserName

ClSOOClF

CAS RN 351003-49-1

IUPAC Name: 5-chloro-2-fluorobenzene-1-sulfonyl chloride
Synonyms: 5-chloro-2-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 229.05
Molecular Formula: C6H3Cl2FO2S
InChi Key: OZKAHSNNKADHTK-UHFFFAOYSA-N
SMILES: FC1=C(C=C(Cl)C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results