Learn More
5-Bromo-2-hydroxypyridine, 97%
Product Code. 11450828
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450828 25 g 25g
11440828 5 g 5g
11460828 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11450828 Supplier Thermo Scientific Alfa Aesar Supplier No. B25267.14
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
CAS: 13466-38-1 | C5H4BrNO | 173.997 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13466-38-1
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 173.997
MDL Number MFCD00234042
InChI Key NDMZZQRNZFWMEZ-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxypyridine, 5-bromopyridin-2-ol, 2-hydroxy-5-bromopyridine, 5-bromopyridin-2 1h-one, 5-bromo-2-pyridinol, 5-bromo-2 1h-pyridinone, 5-bromo-2 1h-pyridone, 2 1h-pyridinone, 5-bromo, 3-bromo-6-hydroxypyridine, 5-bromo-1,2-dihydropyridin-2-one
PubChem CID 599528
IUPAC Name 5-bromo-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1Br
Specifications
Melting Point 178°C to 182°C
Quantity 25 g
Beilstein 108751
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-hydroxypyridine