accessibility menu, dialog, popup

UserName

BrNHO

CAS RN 13466-38-1

IUPAC Name: 5-bromo-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-1H-pyridin-2-one
Molecular Weight (g/mol): 174
Molecular Formula: C5H4BrNO
InChi Key: NDMZZQRNZFWMEZ-UHFFFAOYSA-N
SMILES: BrC1=CNC(=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)