Learn More
5,6-Dihydro-4h-Pyrrolo[3,4-D]Thiazole Dihydrochloride, TRC
Product Code. 30377364
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377364 10 mg 10mg
30371223 50 mg 50mg
30372908 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30377364 Supplier Toronto Research Chemicals Supplier No. TRCD67959310MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1393686-96-8
Molecular Formula C5H6N2S.2HCl
Molecular Weight (g/mol) 199.1
Synonym 4H-Pyrrolo[3,4-d]thiazole, 5,6-dihydro-, hydrochloride (1:2) (ACI), 5,6-Dihydro-4H-pyrrolo[3,4-d]thiazole dihydrochloride
SMILES Cl.Cl.C1NCc2scnc12
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C5H6N2S.2ClH/c1-4-5(2-6-1)8-3-7-4;;/h3,6H,1-2H2;2*1H
Formula Weight 197.9785
Chemical Name or Material 5,6-Dihydro-4h-Pyrrolo[3,4-D]Thiazole Dihydrochloride