CAS RN 1393686-96-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5,6-Dihydro-4h-Pyrrolo[3,4-D]Thiazole Dihydrochloride, TRC
CAS: 1393686-96-8 Molecular Formula: C5H6N2S.2HCl Molecular Weight (g/mol): 199.1 Synonym: 4H-Pyrrolo[3,4-d]thiazole, 5,6-dihydro-, hydrochloride (1:2) (ACI),5,6-Dihydro-4H-pyrrolo[3,4-d]thiazole dihydrochloride SMILES: Cl.Cl.C1NCc2scnc12