Learn More
5-(4-Bromophenyl)-1H-tetrazole, 97%, Thermo Scientific™
Product Code. 11388186
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388186 250 mg 250mg
11378186 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11388186 Supplier Thermo Scientific Alfa Aesar Supplier No. L15181.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 50907-23-8
Molecular Formula C7H5BrN4
Molecular Weight (g/mol) 225.049
MDL Number MFCD00991874
InChI Key YMZMDMWMKVPGDP-UHFFFAOYSA-N
Synonym 5-4-bromophenyl-1h-tetrazole, 5-4-bromo-phenyl-2h-tetrazole, 5-4-bromophenyl-2h-tetrazole, 5-4-bromophenyl tetrazole, 1h-tetrazole, 5-4-bromophenyl, 5-4-bromophenyl-1h-1,2,3,4-tetrazole, 5-4-bromophenyl-2h-1,2,3,4-tetrazole, 5-4-bromophenyl-1h-1,2,3,4-tetraazole, acmc-20aiyk
PubChem CID 766141
IUPAC Name 5-(4-bromophenyl)-2H-tetrazole
SMILES C1=CC(=CC=C1C2=NNN=N2)Br
Spécification
Melting Point ∽268°C (decomposition)
Quantity 250 mg
Beilstein 141235
Formula Weight 225.05
Percent Purity 97%
Chemical Name or Material 5-(4-Bromophenyl)-1H-tetrazole
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.