accessibility menu, dialog, popup

UserName

BrNNNN

CAS RN 50907-23-8

IUPAC Name: 5-(4-bromophenyl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(4-bromophenyl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 224.04
Molecular Formula: C7H4BrN4
InChi Key: UWBMZNMSGITDRY-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=N[N-]N=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results