Learn More
5-(2-Fluorophenyl)-1H-tetrazole, 95%, Thermo Scientific™
Product Code. 11334136
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334136 250 mg 250mg
11324136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11334136 Supplier Thermo Scientific Alfa Aesar Supplier No. L19259.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50907-19-2
Molecular Formula C7H5FN4
Molecular Weight (g/mol) 164.143
MDL Number MFCD00798373
InChI Key ZRFGTMYIGSTCSZ-UHFFFAOYSA-N
Synonym 5-2-fluorophenyl-1h-tetrazole, 5-2-fluorophenyl tetrazole, 5-2-fluorophenyl-2h-tetrazole, 5-2-fluorophenyl-1h-1,2,3,4-tetrazole, 1h-tetrazole, 5-2-fluorophenyl, acmc-20am9w, 2-fluorophenyl tetrazole, maybridge1_000232, 5-2-fluorophenyl-1h-tetraazole, 5-2-fluoro-phenyl-1h-tetrazole
PubChem CID 596275
IUPAC Name 5-(2-fluorophenyl)-2H-tetrazole
SMILES C1=CC=C(C(=C1)C2=NNN=N2)F
Specifications
Melting Point 157°C to 159°C
Quantity 250 mg
Beilstein 1105868
Formula Weight 164.14
Percent Purity 95%
Chemical Name or Material 5-(2-Fluorophenyl)-1H-tetrazole