accessibility menu, dialog, popup

UserName

NNHNNF

CAS RN 50907-19-2

IUPAC Name: 5-(2-fluorophenyl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(2-fluorophenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 164.14
Molecular Formula: C7H5FN4
InChi Key: ZRFGTMYIGSTCSZ-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C1=NNN=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results