Learn More
Tocris Bioscience™ (RS)-3,5-DHPG
Description
(RS)-3,5-DHPG is a selective group I metabotropic glutamate receptor agonist which activates both mGlu1 and mGlu5. Also reported to be an antagonist at metabotropic glutamate receptors linked to phospholipase D.
S-enantiomer also available.
Specifications
Specifications
| Chemical Name or Material | (RS)-3,5-Dihydroxyphenylglycine |
| CAS | 19641-83-9 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Solubility | Soluble to 10mM in water |
| Molecular Formula | C8H9NO4 |
| Formula Weight | Observed MW: 183.16 |
For Research Use Only