missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzonitrile, 98%, Thermo Scientific™

Product Code. 11474474
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474474 5 g 5g
11464474 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474474 Supplier Thermo Scientific Alfa Aesar Supplier No. A13124.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332-25-2
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD00039474
InChI Key XWHIXOMWXCHJPP-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzonitrile, benzonitrile, 4-trifluoromethoxy, p-trifluoromethoxybenzonitrile, p-cyanotrifluoromethoxybenzene, p-trifluoromethoxy benzonitrile, 1-cyano-4-trifluoromethoxy benzene, ncr doxfff, 4-trifluoromethoxy benzenecarbonitrile, pubchem4826, acmc-1coco
PubChem CID 67621
IUPAC Name 4-(trifluoromethoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC(F)(F)F

Specifications

Density 1.285
Boiling Point 195°C to 196°C
Flash Point 82°C (179°F)
Refractive Index 1.452
Quantity 5 g
UN Number UN3276
Beilstein 2832939
Formula Weight 187.12
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.