CAS RN 332-25-2
CAS RN 332-25-2
IUPAC Name:
4-(trifluoromethoxy)benzonitrile
Synonyms:
4-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol):
187.12
Molecular Formula:
C8H4F3NO
InChi Key:
XWHIXOMWXCHJPP-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-(Trifluoromethoxy)benzonitrile, 98%, Thermo Scientific™
CAS: 332-25-2 Molecular Formula: C8H4F3NO Molecular Weight (g/mol): 187.121 MDL Number: MFCD00039474 InChI Key: XWHIXOMWXCHJPP-UHFFFAOYSA-N Synonym: 4-trifluoromethoxy benzonitrile,benzonitrile, 4-trifluoromethoxy,p-trifluoromethoxybenzonitrile,p-cyanotrifluoromethoxybenzene,p-trifluoromethoxy benzonitrile,1-cyano-4-trifluoromethoxy benzene,ncr doxfff,4-trifluoromethoxy benzenecarbonitrile,pubchem4826,acmc-1coco PubChem CID: 67621 IUPAC Name: 4-(trifluoromethoxy)benzonitrile SMILES: C1=CC(=CC=C1C#N)OC(F)(F)F