Learn More
4-Nitrophenylglyoxal hydrate, 97%, Thermo Scientific™
Product Code. 15427457
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427457 1 g 1g
15437457 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15427457 Supplier Thermo Scientific Alfa Aesar Supplier No. H35009.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
As important industrial compounds and are mainly used as solvents. A new method for arginase assay
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4974-57-6
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00038682
InChI Key ILVKBBGIGNSVDO-UHFFFAOYSA-N
Synonym 4-nitrophenylglyoxal, p-nitrophenylglyoxal, 2-4-nitrophenyl-2-oxoacetaldehyde, para-nitrophenylglyoxal, 4-nitrophenyl glyoxal, 1-4-nitrophenyl glyoxal, 4-nitrophenyl oxo acetaldehyde, 2-4-nitrophenyl-2-oxo-acetaldehyde, benzeneacetaldehyde, 4-nitro-alpha-oxo
PubChem CID 151242
IUPAC Name 2-(4-nitrophenyl)-2-oxoacetaldehyde
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(=O)C=O
Specifications
Melting Point 96°C to 98°C
Color Cream
Quantity 1 g
Formula Weight 179.13 (anhy.)
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylglyoxal hydrate