accessibility menu, dialog, popup

UserName

CAS RN 4974-57-6

ONOOO

CAS RN 4974-57-6

IUPAC Name: 2-(4-nitrophenyl)-2-oxoacetaldehyde
Synonyms: 2-(4-nitrophenyl)-2-oxoacetaldehyde
Molecular Weight (g/mol): 179.13
Molecular Formula: C8H5NO4
InChi Key: ILVKBBGIGNSVDO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)C=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results