Learn More
Melford 4-Nitrophenyl-a-L-fucopyranoside
Product Code. p-200087218
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. Quantity unitSize
30018556 1 g 1g
30018555 100 mg 100mg
2 options
Product Code. 30018556
Brand: Melford N120501.0
Chromogenic substrate for detecting a-L-Fucosidase.
Chromogenic substrate for detecting a-L-Fucosidase.
Specifications
Quantity 1 g