Learn More
Melford 4-Nitrophenyl-a-D-mannopyranoside
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Description
Spectrophotometric substrate for detection of α-Mannosidase.
Spectrophotometric substrate for detection of α-Mannosidase.
Free PNP: <200ppm
Specifications
Specifications
| Quantity | 500 mg |