Learn More
Melford 4-Nitrophenyl-a-D-mannopyranoside
Product Code. 30018620
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Product Code. 30018620
Brand: Melford N810600.5
Spectrophotometric substrate for detection of α-Mannosidase.
Spectrophotometric substrate for detection of α-Mannosidase.
Free PNP: <200ppm
Specifications
Quantity 500 mg