missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylbenzoyl chloride, 98%, Thermo Scientific™

Product Code. 11495513
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495513 1 g 1g
11405523 5 g 5g
11415523 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495513 Supplier Thermo Scientific Alfa Aesar Supplier No. A13452.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It employed as an intermediate for pharmaceutical.

Solubility
Reacts with water. Soluble in hot hexane, toluene, dichloromethane.

Notes
Moisture Sensitive, store way form water/moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52710-27-7
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.65
MDL Number MFCD00000698
InChI Key NZYPCJXREKMMCJ-UHFFFAOYSA-N
Synonym p-propylbenzoyl chloride, 4-n-propylbenzoyl chloride, benzoyl chloride, 4-propyl, 4-propylbenzoylchloride, 4-n-propylbenzoylchloride, 4-propyl benzoyl chloride, 4-propyl-benzoyl chloride, 4-n-propyl benzoyl chloride, nzypcjxrekmmcj-uhfffaoysa, 4-propyl-1-benzenecarbonyl chloride
PubChem CID 104293
IUPAC Name 4-propylbenzoyl chloride
SMILES CCCC1=CC=C(C=C1)C(Cl)=O

Specifikationer

Boiling Point 124°C to 126°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3265
Beilstein 2206452
Formula Weight 182.65
Percent Purity 98%
Chemical Name or Material 4-n-Propylbenzoyl chloride

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.