accessibility menu, dialog, popup

UserName

CH3ClO

CAS RN 52710-27-7

IUPAC Name: 4-propylbenzoyl chloride
Synonyms: 4-propylbenzoyl chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: NZYPCJXREKMMCJ-UHFFFAOYSA-N
SMILES: CCCC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)