Learn More
4-n-Pentylphenyl 4-n-propylbenzoate, 97%, Thermo Scientific™
Product Code. 15481518
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481518 1 g 1g
15491518 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15481518 Supplier Thermo Scientific Alfa Aesar Supplier No. H55306.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50649-60-0
Molecular Formula C21H26O2
Molecular Weight (g/mol) 310.437
MDL Number MFCD00041941
InChI Key WNBFPAKRCJNBBS-UHFFFAOYSA-N
Synonym benzoic acid, 4-propyl-, 4-pentylphenyl ester, 4-pentylphenyl-4'-propylbenzoate, 4-pentylphenyl 4-propylbenzoate, 4-n-propyl benzoic acid-4'-n-pentyl phenol ester, p-pentyl phenyl-p-propylbenzoate, 4-n-pentylphenyl 4-n-propylbenzoate, 4-propylbenzoic acid 4-pentylphenyl ester
PubChem CID 170879
IUPAC Name (4-pentylphenyl) 4-propylbenzoate
SMILES CCCCCC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)CCC
Specifications
Melting Point 15°C to 18°C
Density 0.989
Refractive Index 1.543
Quantity 1 g
Formula Weight 310.44
Percent Purity 97%
Chemical Name or Material 4-n-Pentylphenyl 4-n-propylbenzoate