accessibility menu, dialog, popup

UserName

CAS RN 50649-60-0

H3COOCH3

CAS RN 50649-60-0

IUPAC Name: 4-pentylphenyl 4-propylbenzoate
Synonyms: 4-pentylphenyl 4-propylbenzoate
Molecular Weight (g/mol): 310.44
Molecular Formula: C21H26O2
InChi Key: WNBFPAKRCJNBBS-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(OC(=O)C2=CC=C(CCC)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results