Learn More
4-n-Nonylaniline, 98%
Product Code. 15436227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436227 1 g 1g
15446227 5 g 5g
15456227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15436227 Supplier Thermo Scientific Alfa Aesar Supplier No. H33414.03
Request Bulk or Custom Format
CAS: 37529-29-6 | C15H25N | 219.37 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 37529-29-6
Molecular Formula C15H25N
Molecular Weight (g/mol) 219.37
MDL Number MFCD00038370
InChI Key FDECURPHVMNAKO-UHFFFAOYSA-N
Synonym p-nonylaniline, p-n-nonylaniline, 4-n-nonylaniline, p-nonyl aniline, p-nonylamiline, benzenamine, 4-nonyl, 1-amino-4-n-nonylbenzene
PubChem CID 142170
IUPAC Name 4-nonylaniline
SMILES CCCCCCCCCC1=CC=C(N)C=C1
Specifications
Boiling Point 144°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 219.37
Percent Purity 98%
Chemical Name or Material 4-n-Nonylaniline