accessibility menu, dialog, popup

UserName

CAS RN 37529-29-6

H3CNH2

CAS RN 37529-29-6

IUPAC Name: 4-nonylaniline
Synonyms: 4-nonylaniline
Molecular Weight (g/mol): 219.37
Molecular Formula: C15H25N
InChi Key: FDECURPHVMNAKO-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CC=C(N)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)