Learn More
4-(Methylthio)butylnitrile, TRC
Product Code. 30481124
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481124 50 mg 50mg
30475900 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30481124 Supplier Toronto Research Chemicals Supplier No. TRCM33070050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 59121-24-3
Molecular Formula C5 H9 N S
Molecular Weight (g/mol) 115.2
IUPAC Name 4-methylsulfanylbutanenitrile
SMILES CSCCCC#N
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C5H9NS/c1-7-5-3-2-4-6/h2-3,5H2,1H3
Formula Weight 115.0456
Chemical Name or Material 4-(Methylthio)butylnitrile