CAS RN 59121-24-3
CAS RN 59121-24-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(Methylthio)butylnitrile, TRC
CAS: 59121-24-3 Molecular Formula: C5 H9 N S Molecular Weight (g/mol): 115.2 IUPAC Name: 4-methylsulfanylbutanenitrile SMILES: CSCCCC#N