Learn More
4-(Methylsulfonyl)aniline, ≥97%, Thermo Scientific™
Product Code. p-7055970
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422009 25 g 25g
15432009 100 g 100g
15442009 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15422009 Supplier Thermo Scientific Alfa Aesar Supplier No. H64697.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate. A series of 4-(methylsulfonyl)aniline derivatives were synthesized in order to obtain new compounds as a potential anti-inflammatory agents with expected selectivity against COX-2 enzyme.
Solubility
Soluble in water.
Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5470-49-5
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00025353
InChI Key XJEVFFNOMKXBLU-UHFFFAOYSA-N
Synonym 4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
PubChem CID 79617
SMILES CS(=O)(=O)C1=CC=C(N)C=C1
Specifications
Melting Point 133°C to 138°C
Color White
Density 1.44 g/mL
Quantity 25 g
UN Number UN2811
Beilstein 2208425
Sensitivity Light sensitive
Solubility Information Soluble in water.
IUPAC Name 4-methanesulfonylaniline
Formula Weight 171.21
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)aniline