accessibility menu, dialog, popup

UserName

CAS RN 5470-49-5

CH3SOONH2

CAS RN 5470-49-5

IUPAC Name: 4-methanesulfonylaniline
Synonyms: 4-methylsulfonylaniline
Molecular Weight (g/mol): 171.21
Molecular Formula: C7H9NO2S
InChi Key: XJEVFFNOMKXBLU-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(N)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results