missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenoxyacetonitrile, 97%, Thermo Scientific™

Product Code. 11468767
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468767 25 g 25g
11458767 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468767 Supplier Thermo Scientific Alfa Aesar Supplier No. A19926.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33901-44-9
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00068121
InChI Key GOCOKJDTFHEYKP-UHFFFAOYSA-N
Synonym 4-methylphenoxyacetonitrile, 2-4-methylphenoxy acetonitrile, 2-4-methylphenoxy ethanenitrile, p-tolyloxyacetonitrile, p-tolyloxy-acetonitrile, 2-p-tolyloxy acetonitrile, acmc-1ad12, 4-methylphenoxy acetonitrile, p-methylphenoxy-acetonitrile, gocokjdtfheykp-uhfffaoysa
PubChem CID 520657
IUPAC Name 2-(4-methylphenoxy)acetonitrile
SMILES CC1=CC=C(OCC#N)C=C1

Specifications

Melting Point 42°C to 44°C
Boiling Point 76°C to 78°C (11 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 2438642
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 4-Methylphenoxyacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.