accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 33901-44-9

IUPAC Name: 2-(4-methylphenoxy)acetonitrile
Synonyms: 2-(4-methylphenoxy)acetonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: GOCOKJDTFHEYKP-UHFFFAOYSA-N
SMILES: CC1=CC=C(OCC#N)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results