missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15498025
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498025 250 mg 250mg
15488025 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498025 Supplier Thermo Scientific Alfa Aesar Supplier No. H32242.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694514-21-1
Molecular Formula C8H6ClF3O3S
Molecular Weight (g/mol) 274.638
MDL Number MFCD07774167
InChI Key AXNBCYIRTWOTHS-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl benzenesulphonyl chloride, 4-methoxy-3-trifluoromethyl benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl
PubChem CID 22482384
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)Cl)C(F)(F)F

Specifications

Melting Point 52°C to 55°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 274.64
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.