accessibility menu, dialog, popup

UserName

H3COSOOClFFF

CAS RN 694514-21-1

IUPAC Name: 4-methoxy-3-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 4-methoxy-3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 274.64
Molecular Formula: C8H6ClF3O3S
InChi Key: AXNBCYIRTWOTHS-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)S(Cl)(=O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results