Learn More
4-Methoxy-2-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™
Product Code. 11491270
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491270 1 g 1g
11401280 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11491270 Supplier Thermo Scientific Alfa Aesar Supplier No. H26361.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 886503-52-2
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.15
MDL Number MFCD06660317
InChI Key NPUIWDQXZDLZIM-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy-4-methoxybenzaldehyde, 4-methoxy-2-trifluoromethoxy benzaldehyde, 4-formyl-3-trifluoromethoxy anisole, 4-methoxy-2-trifluoromethyloxy benzaldehyde, 2-formyl-5-methoxy-alpha,alpha,alpha-trifluoroanisole
PubChem CID 17750806
IUPAC Name 4-methoxy-2-(trifluoromethoxy)benzaldehyde
SMILES COC1=CC(OC(F)(F)F)=C(C=O)C=C1
Specifications
Refractive Index 1.487
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-2-(trifluoromethoxy)benzaldehyde