accessibility menu, dialog, popup

UserName

CH3OOOFFF

CAS RN 886503-52-2

IUPAC Name: 4-methoxy-2-(trifluoromethoxy)benzaldehyde
Synonyms: 4-methoxy-2-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: NPUIWDQXZDLZIM-UHFFFAOYSA-N
SMILES: COC1=CC(OC(F)(F)F)=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results