Learn More
4-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™
Product Code. 10502865
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10502865 1 g 1g
10235334 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10502865 Supplier Thermo Scientific Maybridge Supplier No. CC45806DA
 Restricted Item
Estimated Shipment: 23-12-2026
Chemical Identifiers
CAS 887922-92-1
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD08690262
InChI Key TVGVHXVLMROUII-UHFFFAOYSA-N
Synonym 4-isocyanato-1-methyl-1h-indole, 1h-indole, 4-isocyanato-1-methyl, 1-methylindol-4-isocyanate, 1h-indole,4-isocyanato-1-methyl, 1-methyl-1h-indole-4-yl isocyanate
PubChem CID 18525783
IUPAC Name 4-isocyanato-1-methylindole
SMILES CN1C=CC2=C(C=CC=C21)N=C=O
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 4-Isocyanato-1-methyl-1H-indole