accessibility menu, dialog, popup

UserName

CAS RN 887922-92-1

IUPAC Name: 4-isocyanato-1-methyl-1H-indole
Synonyms: 4-isocyanato-1-methylindole
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: TVGVHXVLMROUII-UHFFFAOYSA-N
SMILES: CN1C=CC2=C(C=CC=C12)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results