missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxymelatonin, TRC

Product Code. 30136114
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136114 25 mg 25mg
30010953 1 mg 1mg
30011291 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136114 Supplier Toronto Research Chemicals Supplier No. TRCH94463525MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39998-64-6
Molecular Formula C13H16N2O3
Molecular Weight (g/mol) 248.28
Synonym N-[2-(4-hydroxy-5-methoxy-1H-indol-3-yl)ethyl]acetamide
SMILES CC(NCCC1=CNC2=C1C(O)=C(OC)C=C2)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C13H16N2O3/c1-8(16)14-6-5-9-7-15-10-3-4-11(18-2)13(17)12(9)10/h3-4,7,15,17H,5-6H2,1-2H3,(H,14,16)
Formula Weight 248.1161
Percent Purity >95%
Chemical Name or Material 4-Hydroxymelatonin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.